Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Alkaline Phosphatase, Human Placenta

Catalog No. 524604100U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-460-4100U 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-460-4100U Supplier MilliporeSigma™ Supplier No. 524604100U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For life science applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Source Human Placenta
Quantity 100 U
Product Type Alkaline Phosphatase, Human Placenta
Form Liquid

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.