Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set I, Animal-Free

Catalog No. 5351421ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-514-21ML 1 mL
53-514-21SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-514-21ML Supplier MilliporeSigma™ Supplier No. 5351421ML
Add to Cart
Add to Cart

The Protease Inhibitor Cocktail Set I, Animal-Free controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cocktail of five protease inhibitors for the inhibition of a broad range of proteases and esterases. Each vial, when reconstituted with 1mL H2O, yields a 100X stock solution. When diluted to 1X the cocktail contains 500μM AEBSF, HCl (Cat. No. 101500), 150nM Aprotinin (Cat. No.616371), 1μM E-64 (Cat. No. 324890), 0.5mm EDTA, Disodium Salt and 1μM Leupeptin Hemisulfate (Cat. No.108975). Available as 1mL or as a set of 10 x 1 ml.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins A broad range of proteases and esterases
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.