Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PARP Inhibitor VIII, PJ34

Catalog No. 5281501MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-815-01MG 1 mg
52-815-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-815-01MG Supplier MilliporeSigma™ Supplier No. 5281501MG
Add to Cart
Add to Cart

The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins PARP
CAS 344458-15-7
Molecular Formula C17H17N3O2·HCl
Formula Weight 331.8g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.