Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Purmorphamine

Catalog No. 54022020MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
20 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-022-020MG 20 mg
54-022-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-022-020MG Supplier MilliporeSigma™ Supplier No. 54022020MG
Add to Cart
Add to Cart

A cell-permeable activator of Hedgehog signaling that induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 (EC50 = 1 μM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Activation of the Hedgehog signaling pathway
CAS 483367-10-8
Molecular Formula C31H32N6O2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 20 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.