Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ γ-Secretase Inhibitor IX

Catalog No. 56578410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-578-410MG 10 mg
56-578-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-578-410MG Supplier MilliporeSigma™ Supplier No. 56578410MG
Add to Cart
Add to Cart

InSolution™ g-Secretase Inhibitor IX, CAS 208255-80-5, is a 25 mM (5 mg/462 μl or 10 mg/924 μl) solution of g-secretase Inhibitor IX (DAPT; Cat. No. 565770) in DMSO.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Formulation A 25 mM (5mg/462μl or 10mg/924μl) solution of γ-Secretase Inhibitor IX in DMSO.
Target Proteins Aβtotal
Molecular Formula C23H26F2N2O4
Formula Weight 432.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 10 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.