Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Secretase Inhibitor II

Catalog No. 5657491MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-574-91MG 1 mg
56-574-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-574-91MG Supplier MilliporeSigma™ Supplier No. 5657491MG

The β-Secretase Inhibitor II, also referenced under CAS 263563-09-3, controls the biological activity of β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Val-Leu-Leu-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins β-secretase
CAS 263563-09-3
Molecular Formula C25H39N3O5
Formula Weight 461.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.