Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Annexin V-FITC Apoptosis Detection Kit II

Catalog No. CBA0591KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBA0591KIT 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBA0591KIT Supplier MilliporeSigma™ Supplier No. CBA0591KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identifies changes in plasma membrane that occur during apoptosis

  • Highly sensitive, specific fluorometric assay
  • Applications:
    • For measuring Annexin V in apoptotic cells
    • For identification of changes in plasma membrane that occur during apoptosis
    • Used to monitor apoptopic cells using flow cytometry or immunofluorescence microscope
    • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 Tests/Kit
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.