Learn More
Description
Specifications
Specifications
| Color | White |
| Target Proteins | Promotes assembly of microtubules and inhibits tubulins disassembly |
| CAS | 33069-62-4 |
| Molecular Formula | C47H51NO14 |
| Formula Weight | 853.9g/mol |
| Solubility | DMSO or MeOH |
| Content And Storage | Protect from Light, Protect from Moisture |
| Form | Solid |
| Quantity | 100 mg |
| Purity | 97% by HPLC |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.