Learn More
MilliporeSigma™ Calbiochem™ Trypsin Inhibitor, Soybean, High Activity
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Color | White |
| Target Proteins | Trypsin |
| CAS | 9035-81-8 |
| Formula Weight | 20000g/mol |
| Solubility | 67 mM potassium phosphate buffer, pH 7.6 |
| Content And Storage | Protect from moisture |
| Form | Lyophilized |
| Quantity | 100 mg |
| Purity | 90% by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.