Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Forskolin, 7-Deacetyl-7-[O-(N-methylpiperazino)-γ-butyryl]-, Dihydrochloride

Catalog No. 34427310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-427-310MG 10 mg
34-427-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-427-310MG Supplier MilliporeSigma™ Supplier No. 34427310MG
Add to Cart
Add to Cart

Chemically-modified forskolin with greater stability and water solubility than the parent compound.

Chemically-modified forskolin with greater stability and water solubility than the parent compound. Activates adenylate cyclase (EC50 = 13μM). Inhibits 5HT-stimulated inositol phosphate accumulation in smooth muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Adenylate cyclase
CAS 115116-37-5
Molecular Formula C29H48N2O7·2HCl
Formula Weight 609.6g/mol
Solubility MeOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Purity 92% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.