Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Forskolin, Coleus forskohlii

Catalog No. 34427025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-427-025MG 25 mg
34-427-010MG 10 mg
34-427-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34-427-025MG Supplier MilliporeSigma™ Supplier No. 34427025MG
Add to Cart
Add to Cart

Forskolin, CAS 66575-29-9, is a cell-permeable diterpene inotropic agent that acts as an activator of adenylate cyclase (EC50 = 4 mM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins Adenylate cyclase
CAS 66575-29-9
Molecular Formula C22H34O7
Formula Weight 410.5g/mol
Solubility DMSO
Form Crystalline Solid
Quantity 25 mg
Purity 99% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.