Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Glycerophosphate, Disodium Salt, Pentahydrate

Catalog No. 35675250GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-675-250GM 250 g
35-675-100GM 100 g
35-675-50GM 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 35-675-250GM Supplier MilliporeSigma™ Supplier No. 35675250GM
Add to Cart
Add to Cart

A phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor.

Contaminants: L-α-glycerophosphate: ≤5%
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins protein phosphatase
CAS 13408-09-8
Molecular Formula C3H7O6PNa2·5H2O
Formula Weight 306.1g/mol
Solubility H2O
Content And Storage Protect from moisture
Form Solid
Quantity 250 g
Purity 99% by 31P NMR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.