Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cleland’s Reagent

Catalog No. 2331551KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-51KG 1 kg
23-315-510GM 10 g
23-315-525GM 25 g
23-315-5100GM 100 g
23-315-55GM 5 g
23-315-5500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23-315-51KG Supplier MilliporeSigma™ Supplier No. 2331551KG
Add to Cart
Add to Cart

Protective agent for SH groups

  • Cyclizes as it reduces disulfides to thiols, so reaction is "driven" to completion
  • Does not contain waxy surfactant found in many other preparations
  • Oxidized dithiothreitol: ≤0.5%
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 154.24
Purity ≥97% by titration
CAS 12-3-3483
Molecular Formula C4H10O2S2
Content And Storage 2°C to 8°C
Form Powder
Quantity 1 kg
Synonym DTT, Dithiothreitol
Solubility EtOH, Water
Product Type Calbiochem Cleland’s Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.