Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bcr-abl Inhibitor IV, Imatinib

Catalog No. 5045950001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-459-50001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-459-50001 Supplier MilliporeSigma™ Supplier No. 5045950001
Add to Cart
Add to Cart

A cell-permeable, orally bioavailable 2-phenylaminopyrimidine derived compound that competitively binds to ATP-binding site of Abl and stabilizes the inactive conformation, and acts as a highly potent and reversible inhibitor of Abl (IC50 = 38, 25 and 25nM for v-Abl, Bcr-Abl, and c-Abl, respective

A cell-permeable, orally bioavailable 2-phenylaminopyrimidine derived compound that competitively binds to ATP-binding site of Abl and stabilizes the inactive conformation, and acts as a highly potent and reversible inhibitor of Abl (IC50 = 38, 25 and 25nM for v-Abl, Bcr-Abl, and c-Abl, respectively). Also shown to block the activity of PDGFR and c-kit (IC50 = 50 and 100nM). Exhibits high selectivity over IR, IGF-1R, EGFR, c-Src, JAK-2, PKA, PKC isozymes, CKI, and CKII (IC50 >100μM). Preferentially blocks the proliferation of leukemic immature cobblestone-forming area cells and Bcr-abl transfected cell lines, M07/p210 and Ba/F3/p185 (∽1 -10μM), and suppresses tumor growth in mice (50mg/kg, i.p.). Induces apoptosis in Bcr-abl positive cells via intrinsic mitochondrial pathway by down-regulating Bcl-X, promoting cytochrome c release, and activating caspase-9. Does not affect the levels of Bcl-2. Also reported to induce a caspase-independent, necrosis-like programmed cell death mediated by the serine protease activity of Omi/HtrA2.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins Abl
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.