Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bovine γ-Globulin, Serum

Catalog No. 34587625GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-587-625GM 25 g
34-587-65GM 5 g
34-587-6100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34-587-625GM Supplier MilliporeSigma™ Supplier No. 34587625GM
Add to Cart
Add to Cart

Native γ-globulin from bovine serum.

  • Form: Lyophilized
  • Purity: ≥97% by electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
Content And Storage Ok to freeze.
Form Lyophilized
Purity or Quality Grade ≥97% by electrophoresis
Species Bovine serum
Product Type Adult Bovine Serum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.