Learn More
MilliporeSigma™ Calbiochem™ Bovine Serum Albumin, Fraction V (Fatty Acid, Nuclease and Protease Free)
Shop All MilliporeSigma ProductsDescription
- Contaminants
- Ash: ≤3.0%
- Fatty acids: ≤0.02%
- Moisture: ≤5%
- Nuclease: ≤0.1U/mg
- Protease activity: ≤0.1U/mg
- Heavy metals ≤10ppm
- Assay: ≥98% by SDS-PAGE
- CAS: 9048-46-8
- Color: Beige
Recommended Storage
2° to 8°C
Specifications
Specifications
| Quantity | 100 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.