Learn More
MilliporeSigma™ Calbiochem™ Bovine Serum Albumin, Fraction V (Modified Cohn)
Shop All MilliporeSigma ProductsDescription
- Has pH in range of 4.8 to 5.4 in 150mM NaCl (1%)
- Contaminants Moisture: ≤5%
- Heavy metals: ≤50ppm
- Assay: ≥96% by SDS-PAGE
- CAS: 9048-46-8
- Color: Off-White
Recommended Storage
2° to 8°C
Specifications
Specifications
| Quantity | 100 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.