Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, Fraction V, Crystalline

Catalog No. 1265725GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-657-25GM 25 g
12-657-1GM 1 g
12-657-5GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-657-25GM Supplier MilliporeSigma™ Supplier No. 1265725GM
Add to Cart
Add to Cart

Prepared by cold ethanol extraction.

  • Crystalline form
  • Contaminants
  • Moisture: ≤5%
  • Assay: ≥99% by Cellulose Acetate Electrophoresis
  • CAS: 9048-46-8

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.