Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, Fraction V, Fatty Acid-Poor, Endotoxin-Free

Catalog No. 12657910GM Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 g
100 g

Catalog No. 12-657-910GM

Supplier: MilliporeSigma™ 12657910GM

Designed for use in cell culture systems, animal perfusion studies.

  • Fatty acid poor
  • Endotoxin free
  • Contaminants
  • Ash: ≤2.0%
  • Fatty acids: ≤0.2%
  • Moisture: ≤5%
  • Assay: ≥ 95% by Albumin Electrophoresis
  • CAS: 9048-46-8
  • Color: Pale green

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.