Learn More
Description
Includes
Human Recombinant Calcineurin, Bovine Brain Calmodulin, RII Phosphopeptide Substrate, Assay Buffer, Phosphate Detection Reagent, Phosphate Calibration Standard, ½ volume 96-Well Plate, and a user protocol
- Colorimetric assay kit for measuring calcineurin (PP2B) activity and for inhibitor screening
- The assay is performed using RII Phosphopeptide Substrate
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store all components except the 96-Well Plate (room temperature) at -70°C |
| Detection Method | Colorimetric |
| Format | 96-well Plate |
| Sample Type | Purified Human Calcineurin |
| For Use With (Application) | Measuring calcineurin activity |
| Quantity | 1 Kit |
| Synonym | PP2B Assay Kit, Colorimetric |
| Test Time | 3.5h |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
