Learn More
MilliporeSigma™ Calbiochem™ Calcineurin Cellular Activity Assay Kit (Colorimetric)
Catalog No. 2070071KTT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit/ 96 Tests
Catalog No. Quantity
20-700-71KTT 1 Kit/ 96 Tests
1 options
Catalog No. 20-700-71KTT Supplier MilliporeSigma™ Supplier No. 2070071KTT
Add to Cart
Add to Cart
Used for measuring cellular calcineurin (PP2B) enzyme activity.
EMD Millipore Calbiochem™ Calcineurin Cellular Activity Assay Kit (Colorimetric) has convenient 96-well plate format to perform assays.
  • RII Phosphopeptide Substrate supplied with this kit
  • Free phosphate released is detected using malachite green
  • Human recombinant calcineurin is included as positive control
  • Non-radioactive, convenient one-step detection, good sensitivity
  • Packaging: Glass Bottle
Recommended Storage
−70°C
TRUSTED_SUSTAINABILITY
Specifications
Description Assay time: 7 hr
Quantity 1 Kit/ 96 Tests