Learn More
MilliporeSigma™ Calbiochem™ Caspase Detection Kit (Red-VAD-FMK)
Catalog No. QIA921KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit (100 tests)
Catalog No. Quantity
QIA921KIT 1 Kit (100 tests)
1 options
Catalog No. QIA921KIT Supplier MilliporeSigma™ Supplier No. QIA921KIT
For detection of activated caspases in living cells
  • Cell-permeable, non-toxic inhibitor that binds irreversibly to activated caspases in apoptotic cells
  • Can be measured by fluorescence microscopy, fluorescence plate reader, or flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Quantity 1 Kit (100 tests)
Product Type Caspase Detection Kit (Red-VAD-FMK)