Learn More
Specifications
Specifications
| Formulation | Salt-free |
| Quantity | 1 mg |
| Source | Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. |
| Product Type | Collagen, Human Skin Type I |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.