Learn More
MilliporeSigma™ Calbiochem™ Deoxyribonucleic Acid, Sodium Salt, Salmon Testes
Shop All MilliporeSigma ProductsDescription
- Purified DNA from salmon testes
- N/P ratio: 1.5-1.8
- Absorption (0.001% solution in phosphate buffer, pH 7.0, 260 nm): ≥0.155
Specifications
Specifications
| Buffer | Phosphate Buffer |
| CAS | 9007-49-2 |
| Color | Off-white |
| Content And Storage | Store at +2°C to +8°C |
| pH | 7.0 |
| Quantity | 1 g |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.