Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Elastase Inhibitor I

Catalog No. 3246921MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-469-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-469-21MG Supplier MilliporeSigma™ Supplier No. 3246921MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

The Elastase Inhibitor I controls the biological activity of Elastase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Peptide Sequence: Boc-Ala-Ala-Ala-NHO-Bz
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >90% by HPLC
Target Proteins Porcine pancreatic elastase (PPE)
Molecular Formula C21H30N4O7
Formula Weight 450.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.