Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Elastase Substrate IV, Colorimetric

Catalog No. 3246995MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-469-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-469-95MG Supplier MilliporeSigma™ Supplier No. 3246995MG
Add to Cart
Add to Cart

A specific substrate for pancreatic elastase.

Peptide Sequence: Suc-Ala-Ala-Pro-Abu-pNA (Abu = L-α-Aminobutyric Acid)
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Formulation 45mg D-mannitol and 5mg substrate. Sold on the basis of substrate content.
Description 95% by HPLC
Target Proteins A specific substrate for pancreatic elastase
Molecular Formula C25H34N6O9
Formula Weight 562.6g/mol
Solubility EtOH or H2O
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Moisture
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.