Learn More
Description
- Native GDNF (glial derived neurotrophic factor) exists as disulfide homodimeric glycoprotein
- Titrate protein for optimal results in individual systems
Specifications
Specifications
| Formulation | Lyophilized from 35% Acetonitrile, 0.1% TFA, 50μg BSA/μg GDNF |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Common Name | rrGDNF, Glial Derived Neurotrophic Factor |
| Product Type | GDNF, Rat, Recombinant, S. frugiperda |
| Form | Lyophilized |
| Purity or Quality Grade | ≥97% by SDS-PAGE |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.