Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GP Antagonist-2A

Catalog No. 3717801MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-178-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-178-01MG Supplier MilliporeSigma™ Supplier No. 3717801MG
Add to Cart
Add to Cart

The GP Antagonist-2A controls the biological activity of GP. This small molecule/inhibitor is primarily used for Neuroscience applications.

Peptide Sequence: H-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-D-Trp-Met-NH2
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Inhibits the activation of Gq by M1-muscarinic cholinergic receptors
Molecular Formula C79H105N21O13S
Formula Weight 1588.9g/mol
Solubility H2O
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.