Learn More
Description
EMD Millipore Calbiochem™ Human Plasma Fibronectin is purified from human plasma.
- Native fibronectin
- Contaminants: None
- Detected by IEP with antiserum to fibronectin or normal human serum
- Single Major B by SDS-PAGE
- CAS: 86088-83-7
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Human Plasma Fibronectin |
| Formulation | In PBS, pH 7.3. |
| Quantity | 1 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.