Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ I-BET

Catalog No. 4010105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-101-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-101-05MG Supplier MilliporeSigma™ Supplier No. 4010105MG
Add to Cart
Add to Cart

A cell-permeable benzodiazepine compound that binds the tandem bromodomains of BET (bromodomain and extra terminal domain) family members BRD2 (1-473), BRD3 (1-434), and BRD4 (1-477) with high affinity.

A cell-permeable benzodiazepine compound that binds the tandem bromodomains of BET (bromodomain and extra terminal domain) family members BRD2 (1-473), BRD3 (1-434), and BRD4 (1-477) with high affinity ( K d = 61.3, 50.5, and 55.2 nM, respectively, by ITC) and effectively competes against tetra-acetylated H4 peptide (Millipore cat. no. 12-379) for BRD2/3/4 binding (IC50 = 32.5, 42,4, and 36.1 nM, respectively, in competitive equilibrium binding assays), while exhibiting little affinity toward 5 other bromodomain-containing proteins (ATAD2, BAZ2B, CREBBP, PCAF, SP140) or a panel of 38 cellular enzymes, GPCRs, transporters, and ion channels. Shown to differentially modulate LPS-induced gene expression, notably suppressing LPS-induced upregulation of genes involved in inflammatory response, in murine BMDMs (bone marrow-derived macrophages) in vitro (30 min 1μM pretreatment) and effectively prevent LPS-, heat-killed Salmonella typhimurium -, and CLP- (caecal ligation and puncture) induced death in mice in vivo (30mg/kg i.v.). Cellular Brd2/3/4 knockdown using siRNA results in mostly the same effect as I-BET treatment in modulating LPS gene induction profile in murine BMDMs, but not without exceptions, indicating that not all BET-dependent transcription regulations are mediated via its binding to acetylated histones. Also available as a 50mm solution in DMSO (Cat. No. 506071).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish White
Description 98% by Chiral HPLC
CAS 1260907-17-2
Molecular Formula C22H22ClN5O2
Formula Weight 423.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.