Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InnoZyme™ Cathepsin B Activity Assay Kit, Fluorogenic

Catalog No. CBA0011KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
96 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBA0011KIT 96 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBA0011KIT Supplier MilliporeSigma™ Supplier No. CBA0011KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for measurement of cathepsin B activity

  • Sensitive fluorogenic assay
  • Excitation max: 360 to 380nm; Emission max: 430 to 460nm)
  • Cysteine proteinase
  • Implicated in inflammatory airway disease
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Quantity 96 Tests/Kit
Content And Storage -20°C and -70°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.