Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ γ-Secretase Inhibitor X

Catalog No. 565771250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-577-1250UG 250 μg
56-577-1500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-577-1250UG Supplier MilliporeSigma™ Supplier No. 565771250UG
Add to Cart
Add to Cart

InSolution™ γ-Secretase Inhibitor X is a 1 mM solution in DMSO. A highly potent inhibitor of g-secretase (IC50 = 17, 48, 67 nM for Aβtotal, Aβ40 & Aβ42 in SHSY5Y cells overexpressing spβA4CTF).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1 mM (250μg in 372μl or 500μg in 744μl) solution in DMSO.
Target Proteins Aβtotal
Molecular Formula C39H52N4O6
Formula Weight 672.9g/mol
Content And Storage Protect from Light, Protect from Moisture
Form Liquid
Quantity 250 μg
Purity 90% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.