Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ JHDM Inhibitor VIII, SD70

Catalog No. 5316620001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-166-20001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-166-20001 Supplier MilliporeSigma™ Supplier No. 5316620001
Add to Cart
Add to Cart

A cell-permeable, bioavailable, non-toxic 8-hydroxyquinolinyl based compound that binds to and colocalizes with androgen receptor (AR) at both ligand-dependent and independent transcriptional regulatory elements

A cell-permeable, bioavailable, non-toxic 8-hydroxyquinolinyl based compound that binds to and colocalizes with androgen receptor (AR) at both ligand-dependent and independent transcriptional regulatory elements. Acts as an inhibitor of dihydotestosterone-regulated gene expression. Shown to inhibit prostate cancer cell transcription by blocking the activity of histone lysine demethylase KDM4C (IC50 = 30μM). Increases total H3K9me2 levels in 293 T cells in enhancer and promoter regions and causes a slight diminution in the levels of methylated H3K36. Does not affect the activity of DNA/RNA polymerase or topoisomerase I and II activities in any significant manner. Blocks the proliferation of hormone-independent CWR22Rv1 cells in culture (∽ 5μM) and also inhibits their growth in a prostate cancer xenograft murine model (10mg/kg, i.p.daily).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
Target Proteins JMJD2C/KDM4C
Molecular Formula C18H18N2O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.