Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ketoconazole

Catalog No. 42060050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-060-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-060-050MG Supplier MilliporeSigma™ Supplier No. 42060050MG
Add to Cart
Add to Cart

Antifungal agent that displays potent anti-metastatic, anti-neoplastic, and anti-psoriatic activities

  • Inhibitor of cytochrome P-450 in steroid biosynthesis
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Content And Storage 2°C to 8°C
Quantity 50 mg
Product Type Ketoconazole
Form Solid

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.