Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L-CCG-I

Catalog No. 5044670001 Shop All MilliporeSigma Products
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-446-70001 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-446-70001 Supplier MilliporeSigma™ Supplier No. 5.04467.0001

A potent agonist for group II metabotropic glutamate receptors (EC50

A potent agonist for group II metabotropic glutamate receptors (EC50 = 0.49μM and 0.44μM for mGlu2 and mGlu3, respectively). Higher level of activity than glutamate or (±)-trans-ACPD. Widely used in studies related to neurotransmissions mediated via group II mGlu receptors.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins mGlu2 & mGlu3
Molecular Formula C6H9NO4
Solubility Aqueous basic solutions
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.