Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Lipid Hydroperoxide (LPO) Assay Kit

Catalog No. 4376391KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-763-91KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-763-91KIT Supplier MilliporeSigma™ Supplier No. 4376391KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

FTS Reagent 1, FTS Reagent 2, Lipid Hydroperoxide Standard, Extract R, Triphenylphosphine, and user protocol

Used for measurement of hydroperoxides.

EMD Millipore Calbiochem™ Lipid Hydroperoxide (LPO) Assay Kit is sensitive, reliable helps to quantify lipid peroxidation.

    Utilizes redox reactions with ferrous ions
  • Applications: Pathophysiological disorders
  • Format: Cuvette or 96 well plate
  • TotalAssayTime: 1.5 hr
  • Sufficient for 100 Tests
  • Assay Range: 0.25 to 5nmol hydroperoxide per assay tube
  • Sample Type: Tissues, cultured cells, plant materials, foods, biological fluids
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Quantity 1 Kit
For Use With (Application) Measure lipid hydroperoxides
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.