Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ 20X SSPE Buffer Liquid Concentrate

Catalog No. 84104L Shop All MilliporeSigma Products
Catalog No. 84-104-L Supplier MilliporeSigma™ Supplier No. 84104L
Add to Cart
Add to Cart

For use in molecular biology applications

  • Molecular Biology grade: D
  • pH of solution at 25°C: 6.85 to 7.15
  • Protease: None detected
  • RNase: None detected
  • Packaging: Plastic Receptable (Fibreboard boxes)
  • Color: Colorless
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.