Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ DNTP Mixture, 100mM Solution

Catalog No. 36401PC Shop All MilliporeSigma Products
Change view
Click to view available options
DNTP mixture, 100mM Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
36-401-PC DNTP mixture, 100mM Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-401-PC Supplier MilliporeSigma™ Supplier No. 36401PC

Used for hydrolysis of DNA for critical RNA isolation applications.

  • Reconstitute enzyme to concentration of at least 2mg/mL in 50% Glycerol solution (Cat. No. 4750) with 20mM Tris-HCl, pH 7.5 (Cat. No. 9285) and 1mM MgCl2 (Cat. No. 5985)
  • Activity (Kunitz, 25°C): ≥1800U/mg
  • RNase: None detected
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C
Product Type DNTP Mixture, 100mM Solution
Quantity 1

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.