Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ Guanidine Hydrochloride, Molecular Biology Grade
Shop All MilliporeSigma ProductsDescription
EMD Millipore Calbiochem™ OmniPur™ Guanidine Hydrochloride, Molecular Biology Grade is denaturant which inhibits RNase protease activity.
- Specifications:
- A260 (0.1M, water): ≤0.05 AU
- A280 (0.1M, water): ≤0.01 AU
- DNase: None Detected
- Heavy metals (as Pb): ≤0.001%
- Phosphatase: None Detected
- Protease: None Detected
- RNase: None Detected
- Assay: ≥99.5%
- CAS: 50-01-1
- Molecular Formula: H2NC(NH)NH2·HCl
- Color: White
Specifications
Specifications
| Quantity | 10kg |
| Product Type | Guanidine Hydrochloride |
| Content And Storage | 15° to 30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.