Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Guanidine Isothiocyanate, Molecular Biology Grade

Catalog No. 5120250GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-202-50GM 250g
51-401-KG 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51-202-50GM Supplier MilliporeSigma™ Supplier No. 5120250GM
Add to Cart
Add to Cart

Inactivates RNases during isolation of RNA from cell lysate.

EMD Millipore Calbiochem™ OmniPur™ Guanidine Isothiocyanate, Molecular Biology Grade is protein denaturant.

  • Molecular biology grade
  • Specifications:
  • A (280nm, 70%, water): ≤0.8AU
  • Assay: ≥99.0%
  • DNase: None Detected
  • Melting point: 118 to 121°C
  • Protease: None Detected
  • RNase: None Detected
  • Solubility (70%, water): To pass test
  • Assay: ≥99.0%
  • CAS: 593-84-0
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Quantity 250g
Product Type Guanidine Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.