Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ IPTG

Catalog No. 58001GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-001-GM 1 g
58-105-GM 5 g
58-201-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58-001-GM Supplier MilliporeSigma™ Supplier No. 58001GM

Inducer for β-galactosidase, an enzyme that promotes lactose utilization.

EMD Millipore Calbiochem™ OmniPur™ IPTG is used in conjunction with X-Gal.

  • Molecular Biology grade
  • Stock solutions:
  • X-Gal: 20mg/mL in DMF
  • IPTG: 20mg/mL in water
  • Use 4:1 (X-Gal:IPTG)
  • Assay: ≥99%
  • CAS: 367-93-1
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description IPTG, Crystalline Powder
Purity Molecular Biology Grade
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.