Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phospholipase C, Bacillus cereus

Catalog No. 525186250U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-518-6250U 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-518-6250U Supplier MilliporeSigma™ Supplier No. 525186250U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catalyzes hydrolysis of lecithins to 1,2-diglycerides and choline phosphate

TRUSTED_SUSTAINABILITY

Specifications

Color Brownish
Content And Storage -20°C
Source Bacillus cereus
Quantity 250 U
Product Type Phospholipase C
Form Lyophilized Solid

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.