Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PNU-120596

Catalog No. 52820110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-820-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-820-110MG Supplier MilliporeSigma™ Supplier No. 52820110MG
Add to Cart
Add to Cart

A urea derivative that acts as a potent, selective, and reversible positive allosteric modulator for the α7 subtype of neural nicotinic acetylcholine receptors

A urea derivative that acts as a potent, selective, and reversible positive allosteric modulator for the α7 subtype of neural nicotinic acetylcholine receptors (EC50 = 216nM), with no significant effect on α4β2, α3β4 and α9α10 receptors. Specifically increase a-7 nAChR-mediated channel currents and channel mean open time in cultured neurons (∽ 1 mM). However, it does neither affect ion selectivity nor unitary conductance. Shown to cross the blood-brain barrier. Causes conformational changes in the ligand binding domain of a7 nicotinic receptors that partially overlap with those caused by acetylcholine. Causes changes in cysteine accessibility at the inner beta sheet, transition zone, and agonist binding site. Also reported to potentiate agonist-evoked GABAergic postsynaptic inward currents in somatodendritic membrane of hippocampal interneurons.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
CAS 501925-31-1
Molecular Formula C13H14ClN3O4
Formula Weight 311.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.