Learn More
MilliporeSigma™ Calbiochem™ PP2A Inhibitor II, GST-Fusion, Human, Recombinant, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 50 mM Tris-HCl, 14 mM βME, 1 mM benzamidine, 1 mM EDTA, 0.1 mM PMSF, 10% glycerol, pH 7.4. |
| Description | 90% by SDS-PAGE |
| Target Proteins | PP2A |
| Formula Weight | 67000g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles |
| Form | Liquid |
| Quantity | 250 NG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.