Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pronase E

Catalog No. 74332 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-332 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-332 Supplier MilliporeSigma™ Supplier No. 74332

For hydrolysis of glyco protein peptide linkages

  • Molecular biology grade
  • Derived from Streptomyces griseus
  • Protease mixture of exo- and endonucleases

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

TRUSTED_SUSTAINABILITY

Specifications

Color Slightly Brown
Source Streptomyces griseus
Quantity 1 g
Product Type Pronase E
Form Fine Powder

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.