Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein C, Human Plasma

Catalog No. 539215100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-921-5100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-921-5100UG Supplier MilliporeSigma™ Supplier No. 539215100UG
Add to Cart
Add to Cart

Potent anticoagulant that acts through selective inactivation of Factors Va and VIIIa

TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 20mM Tris-HCl, 1mM benzamidine, pH 7.4
Quantity 100 μg
Source Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Storage Requirements ≤ -70°C 
Form Liquid
Purity or Quality Grade ≥95% by SDS-PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.