Learn More
Specifications
Specifications
| Formulation | From 100mM NaCl, 20mM Tris-HCl, 1mM benzamidine, pH 7.4 |
| Quantity | 100 μg |
| Source | Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. |
| Storage Requirements | 2° to 8°C |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by SDS-PAGE |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.