Learn More
Description
- Hematopoietic growth factor that stimulates growth of cells of multiple lineages
Specifications
Specifications
| Formulation | Carrier free |
| Quantity | 10 μg |
| Storage Requirements | ≤-70°C |
| Endotoxin Concentration | ≥0.1ng/μg SCF |
| Common Name | rhSCF |
| Product Type | Recombinant human stem cell factor |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by SDS-PAGE |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.