Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Recombinant Mouse, BMP-4

Catalog No. 20363910UG Shop All MilliporeSigma Products
Change view
Click to view available options
Recombinant Mouse, BMP-4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
20-363-910UG Recombinant Mouse, BMP-4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-363-910UG Supplier MilliporeSigma™ Supplier No. 20363910UG

Expressed in Chinese Hamster Ovary cells

  • Consists of amino acids 293 to 408
  • Plays key role in epidermal determination, the development of multiple organs, suppression of neural induction, tissue repair, and mesenchyme formation
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μm sterile-filtered Acetonitrile, TFA, BSA.
Quantity 10μg
Storage Requirements -20°C
Product Type Recombinant Mouse, BMP-4
Form Liquid
Purity or Quality Grade ≥95% by SDS-PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.