Learn More
Description
- Mouse insulin-like growth factor binding protein 3 (IGFBP-3) consists of amino acids 22-291
- Expressed in NS0-derived murine myeloma cells with four substitutions: Arg250Gln, Gln259Arg, Ser260Gly, Arg271Pro
- Mouse IGFBP-3 cDNA encodes 291 amino acid residue precursor protein with putative 27 aa residue signal peptide
Specifications
Specifications
| Formulation | Lyophilized from 0.2μm filtered Acetonitrile, TFA. |
| Quantity | 25 μg |
| Storage Requirements | -20°C |
| Product Type | Recombinant Mouse IGFBP-3 |
| Form | Solid |
| Purity or Quality Grade | ≥95% by weight |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.